Assay Details
Target Gene Details
Entrez Gene ID: | 7486 |
Gene Name: | Werner syndrome RecQ like helicase |
Gene Aliases: |
RECQ3, RECQL2, RECQL3 |
Location: |
Chr.8:31033262-31173761 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRN | NM_000553.4 | NP_000544.2 | ||
XM_011544639.2 | XP_011542941.1 | |||
AF091214.1 | AAC63361.1 | |||
BP279291.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)