Assay Details
Target Gene Details
Entrez Gene ID: | 55246 |
Gene Name: | coiled-coil domain containing 25 |
Gene Aliases: |
- |
Location: |
Chr.8:27733316-27772656 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC25 | NM_001304529.1 | NP_001291458.1 | ||
NM_001304530.1 | NP_001291459.1 | |||
NM_001304532.1 | NP_001291461.1 | |||
NM_018246.2 | NP_060716.2 | |||
NR_130761.1 | ||||
NR_130762.1 | ||||
XM_011544570.2 | XP_011542872.1 | |||
XM_011544571.2 | XP_011542873.1 | |||
AK001715.1 | BAA91857.1 | |||
AK027900.1 | BAB55442.1 | |||
AK292875.1 | ||||
AK294848.1 | ||||
AK311437.1 | ||||
AK316015.1 | ||||
BC032588.1 | ||||
BC049197.1 | ||||
BC065295.1 | AAH65295.1 | |||
BC110391.1 | ||||
BG718771.1 | ||||
BU602778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv610867 | Chr.8:27607002 - 27757379 on Build GRCh38 | Gain | SCARA3 CCDC25 CLU MIR6843 MIR3622B MIR3622A |
esv3616726 | Chr.8:27741299 - 27744731 on Build GRCh38 | Loss | CCDC25 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |