Assay Details
Target Gene Details
Entrez Gene ID: | 8667 |
Gene Name: | eukaryotic translation initiation factor 3 subunit H |
Gene Aliases: |
EIF3S3, eIF3-gamma, eIF3-p40 |
Location: |
Chr.8:116644816-116755823 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF3H | NM_003756.2 | NP_003747.1 | ||
AK093128.1 | ||||
AK222520.1 | BAD96240.1 | |||
AK222620.1 | BAD96340.1 | |||
AK296162.1 | ||||
AU117206.1 | ||||
BC000386.2 | AAH00386.1 | |||
BC090880.1 | AAH90880.1 | |||
BT019594.1 | AAV38401.1 | |||
CR456906.1 | CAG33187.1 | |||
U54559.1 | AAD03465.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)