Assay Details
Target Gene Details
Entrez Gene ID: | 60487 |
Gene Name: | tRNA methyltransferase 11 homolog |
Gene Aliases: |
C6orf75, MDS024, TRM11, TRMT11-1 |
Location: |
Chr.6:125986430-126083131 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRMT11 | NM_001031712.2 | NP_001026882.2 | ||
XM_017011168.1 | XP_016866657.1 | |||
XM_017011169.1 | XP_016866658.1 | |||
AF182423.1 | AAG14959.1 | |||
AF532977.1 | AAQ10284.1 | |||
AK308651.1 | ||||
BC056878.1 | AAH56878.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1025069 | Chr.6:125973448 - 126003809 on Build GRCh38 | Loss |
![]() ![]() |
nsv1022677 | Chr.6:125935606 - 126195159 on Build GRCh38 | Gain |
![]() ![]() |
nsv1019490 | Chr.6:125927450 - 126221150 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv830796 | Chr.6:125844487 - 126015022 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)