Assay Details
Target Gene Details
Entrez Gene ID: | 135228 |
Gene Name: | CD109 molecule |
Gene Aliases: |
CPAMD7, p180, r150 |
Location: |
Chr.6:73679192-73828317 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD109 | NM_001159587.2 | NP_001153059.1 | ||
NM_001159588.2 | NP_001153060.1 | |||
NM_133493.4 | NP_598000.2 | |||
XM_005248659.3 | XP_005248716.1 | |||
XM_011535473.1 | XP_011533775.1 | |||
AF410459.1 | AAL84159.1 | |||
AK095888.1 | BAC04642.1 | |||
AK313636.1 | ||||
AY149920.1 | AAN78483.1 | |||
AY788891.1 | ||||
BX641095.1 | CAE46045.1 | |||
EF553520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576178 | Chr.6:73593218 - 73711991 on Build GRCh38 | Gain | CD109 LOC101928489 SLC17A5 |
dgv6015n100 | Chr.6:73590773 - 73714632 on Build GRCh38 | Gain | CD109 LOC101928489 SLC17A5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |