Assay Details
Target Gene Details
Entrez Gene ID: | 105378159 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.6:130288003-130386905 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105378159 | XM_017011599.1 | XP_016867088.1 |
Target Gene Details
Entrez Gene ID: | 114801 |
Gene Name: | transmembrane protein 200A |
Gene Aliases: |
KIAA1913, TTMA, TTMC |
Location: |
Chr.6:130366281-130443424 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM200A | NM_001258276.1 | NP_001245205.1 | ||
NM_001258277.1 | NP_001245206.1 | |||
XM_005266816.2 | XP_005266873.1 | |||
AB067500.1 | ||||
AK315824.1 | ||||
BC044246.1 | AAH44246.1 | |||
DA657363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain | L3MBTL3 SAMD3 TMEM244 LOC105378159 LOC102723409 LAMA2 TMEM200A ARHGAP18 |