Assay Details
Target Gene Details
Entrez Gene ID: | 134728 |
Gene Name: | interleukin 1 receptor associated kinase 1 binding protein 1 |
Gene Aliases: |
AIP70, SIMPL |
Location: |
Chr.6:78867486-78979411 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IRAK1BP1 | NM_001010844.3 | NP_001010844.1 | ||
XM_006715336.3 | XP_006715399.1 | |||
XM_011535447.1 | XP_011533749.1 | |||
XM_011535448.2 | XP_011533750.1 | |||
XM_017010264.1 | XP_016865753.1 | |||
XM_017010265.1 | XP_016865754.1 | |||
AK309003.1 | ||||
BC112253.1 | AAI12254.1 | |||
BC112255.1 | AAI12256.1 | |||
CR990080.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |