Assay Details
Target Gene Details
Entrez Gene ID: | 23600 |
Gene Name: | alpha-methylacyl-CoA racemase |
Gene Aliases: |
AMACRD, CBAS4, RACE, RM |
Location: |
Chr.5:33986986-34008115 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMACR | NM_001167595.1 | NP_001161067.1 | ||
NM_014324.5 | NP_055139.4 | |||
NM_203382.2 | NP_976316.1 | |||
AF047020.1 | AAD10205.1 | |||
AF158378.1 | AAF22610.1 | |||
AJ130733.1 | CAB44062.1 | |||
AK000912.1 | ||||
AK022765.1 | ||||
AK222831.1 | BAD96551.1 | |||
AK225749.1 | ||||
AK292988.1 | ||||
AL545355.3 | ||||
AY330488.1 | AAQ67234.1 | |||
AY330489.1 | AAQ94259.1 | |||
AY935981.1 | AAY16192.1 | |||
BC009471.1 | AAH09471.1 | |||
BG708726.1 | ||||
BI550368.1 | ||||
BQ941482.1 | ||||
BT007193.1 | AAP35857.1 | |||
EF560721.1 | ||||
FJ498906.1 | ||||
FJ498907.1 | ||||
FJ498908.1 |
Target Gene Details
Entrez Gene ID: | 100534612 |
Gene Name: | C1QTNF3-AMACR readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.5:33986986-34124528 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1QTNF3-AMACR | NR_037951.1 | |||
AB209261.1 | BAD92498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3604666 | Chr.5:33947465 - 34004885 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv830255 | Chr.5:33929507 - 34109160 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv519240 | Chr.5:33950598 - 34749311 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv522715 | Chr.5:33319157 - 34037875 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)