Assay Details
Target Gene Details
Entrez Gene ID: | 29958 |
Gene Name: | dimethylglycine dehydrogenase |
Gene Aliases: |
DMGDHD, ME2GLYDH |
Location: |
Chr.5:78997564-79069674 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DMGDH | NM_013391.3 | NP_037523.2 | ||
AF111858.1 | AAF21941.1 | |||
AK057641.1 | ||||
AK314736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762527 | Chr.5:78625376 - 79252373 on Build GRCh38 | Gain | ARSB DMGDH LHFPL2 JMY BHMT2 BHMT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |