Assay Details
Target Gene Details
Entrez Gene ID: | 206358 |
Gene Name: | solute carrier family 36 member 1 |
Gene Aliases: |
Dct1, LYAAT1, PAT1, TRAMD3 |
Location: |
Chr.5:151411344-151556318 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC36A1 | XM_011537591.1 | XP_011535893.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3607178 | Chr.5:151420015 - 151420720 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)