Assay Details
Target Gene Details
Entrez Gene ID: | 475 |
Gene Name: | antioxidant 1 copper chaperone |
Gene Aliases: |
ATX1, HAH1 |
Location: |
Chr.5:151742822-151758649 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATOX1 | NM_004045.3 | NP_004036.1 | ||
AK293063.1 | ||||
BC112248.1 | AAI12249.1 | |||
BC112250.1 | AAI12251.1 | |||
BI599233.1 | ||||
BU172675.1 | ||||
CN280769.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |