Assay Details
Target Gene Details
Entrez Gene ID: | 84868 |
Gene Name: | hepatitis A virus cellular receptor 2 |
Gene Aliases: |
CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3 |
Location: |
Chr.5:157085832-157109237 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAVCR2 | NM_032782.4 | NP_116171.3 | ||
AB924452.1 | ||||
AF251707.1 | AAM19100.1 | |||
AF450242.1 | AAL65157.1 | |||
AF450243.1 | AAL65158.1 | |||
AK027334.1 | BAB55044.1 | |||
AK130144.1 | ||||
AK314406.1 | ||||
AY069944.1 | AAL55401.1 | |||
BC063431.1 | AAH63431.1 | |||
JX049979.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1109742 | Chr.5:157099986 - 157110561 on Build GRCh38 | Deletion | HAVCR2 |
esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain | ITK HAVCR1 MED7 HAVCR2 FAM71B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |