Assay Details
Target Gene Details
Entrez Gene ID: | 115123 |
Gene Name: | membrane associated ring-CH-type finger 3 |
Gene Aliases: |
MARCH-III, RNF173 |
Location: |
Chr.5:126867714-127032402 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MARCH3 | NM_178450.4 | NP_848545.1 | ||
XM_011543128.2 | XP_011541430.1 | |||
XM_011543129.1 | XP_011541431.1 | |||
XM_011543130.1 | XP_011541432.1 | |||
XM_011543131.2 | XP_011541433.1 | |||
XM_017009014.1 | XP_016864503.1 | |||
AF055007.1 | ||||
AK290129.1 | ||||
BC047569.1 | AAH47569.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |