Assay Details
Target Gene Details
Entrez Gene ID: | 101928794 |
Gene Name: | uncharacterized LOC101928794 |
Gene Aliases: |
- |
Location: |
Chr.5:67001618-67004154 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928794 | NR_111908.1 | |||
AW003767.1 | ||||
BQ011784.1 |
Target Gene Details
Entrez Gene ID: | 375449 |
Gene Name: | microtubule associated serine/threonine kinase family member 4 |
Gene Aliases: |
- |
Location: |
Chr.5:66596348-67169595 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAST4 | NM_001164664.1 | NP_001158136.1 | ||
NM_001290226.1 | NP_001277155.1 | |||
NM_015183.2 | NP_055998.1 | |||
XM_011543382.2 | XP_011541684.1 | |||
XM_011543385.2 | XP_011541687.1 | |||
XM_017009447.1 | XP_016864936.1 | |||
XM_017009448.1 | XP_016864937.1 | |||
XM_017009449.1 | XP_016864938.1 | |||
XM_017009450.1 | XP_016864939.1 | |||
XM_017009451.1 | XP_016864940.1 | |||
XM_017009452.1 | XP_016864941.1 | |||
AK131421.1 | ||||
AK299538.1 | ||||
AY830839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023631 | Chr.5:66918448 - 67054126 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)