Assay Details
Target Gene Details
Entrez Gene ID: | 408187 |
Gene Name: | serine peptidase inhibitor, Kazal type 14 (putative) |
Gene Aliases: |
SPINK5L2 |
Location: |
Chr.5:148146372-148176076 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPINK14 | XM_017009466.1 | XP_016864955.1 | ||
XM_017009468.1 | XP_016864957.1 | |||
JF509944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3607101 | Chr.5:148020480 - 148213612 on Build GRCh38 | Loss | SPINK5 SPINK6 SPINK14 |
esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss | SPINK9 SPINK13 SPINK5 LOC102546294 SPINK6 LOC105378220 SPINK7 SPINK14 |
nsv830515 | Chr.5:148081289 - 148222684 on Build GRCh38 | Gain+Loss | SPINK5 SPINK6 SPINK14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |