Assay Details
Target Gene Details
Entrez Gene ID: | 2107 |
Gene Name: | eukaryotic translation termination factor 1 |
Gene Aliases: |
D5S1995, ERF, ERF1, RF1, SUP45L1, TB3-1 |
Location: |
Chr.5:138506093-138543300 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ETF1 | NM_001256302.1 | NP_001243231.1 | ||
NM_001282185.1 | NP_001269114.1 | |||
NM_001291974.1 | NP_001278903.1 | |||
NM_001291975.1 | NP_001278904.1 | |||
NM_004730.3 | NP_004721.1 | |||
XM_005271921.1 | XP_005271978.1 | |||
XM_017009221.1 | XP_016864710.1 | |||
XM_017009222.1 | XP_016864711.1 | |||
AK302364.1 | ||||
AK309875.1 | ||||
AK312510.1 | ||||
BC014269.2 | AAH14269.1 | |||
BC088358.1 | AAH88358.1 | |||
BI460904.1 | ||||
BT007374.1 | AAP36038.1 | |||
DC297662.1 | ||||
M75715.1 | AAA36665.1 | |||
U90176.1 | AAB49726.1 | |||
X81625.1 | CAA57281.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |