Assay Details
Target Gene Details
Entrez Gene ID: | 206358 |
Gene Name: | solute carrier family 36 member 1 |
Gene Aliases: |
Dct1, LYAAT1, PAT1, TRAMD3 |
Location: |
Chr.5:151411344-151556318 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC36A1 | XM_011537591.1 | XP_011535893.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |