Assay Details
Target Gene Details
Entrez Gene ID: | 814 |
Gene Name: | calcium/calmodulin dependent protein kinase IV |
Gene Aliases: |
CaMK IV, CaMK-GR, caMK |
Location: |
Chr.5:111223653-111498503 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAMK4 | NM_001323374.1 | NP_001310303.1 | ||
NM_001323375.1 | NP_001310304.1 | |||
NM_001323376.1 | NP_001310305.1 | |||
NM_001323377.1 | NP_001310306.1 | |||
NM_001744.5 | NP_001735.1 | |||
AK311662.1 | ||||
AK312840.1 | ||||
BC016695.1 | AAH16695.2 | |||
BC025687.1 | AAH25687.1 | |||
BG721313.1 | ||||
D30742.1 | BAA06403.1 | |||
DA114495.1 | ||||
DA396146.1 | ||||
L17000.1 | AAA35639.1 | |||
L24959.1 | AAA18251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3606259 | Chr.5:110985051 - 111262481 on Build GRCh38 | Loss | WDR36 CAMK4 TSLP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |