Assay Details
Target Gene Details
Entrez Gene ID: | 30820 |
Gene Name: | potassium voltage-gated channel interacting protein 1 |
Gene Aliases: |
KCHIP1, VABP |
Location: |
Chr.5:170353055-170736632 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNIP1 | NM_001034838.2 | NP_001030010.1 | ||
XM_017009407.1 | XP_016864896.1 | |||
XM_017009408.1 | XP_016864897.1 | |||
AK301775.1 | ||||
DC420716.1 | ||||
DQ148476.1 | AAZ77793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471054 | Chr.5:170278287 - 170532433 on Build GRCh38 | Loss | LINC01366 CTD-2270F17.1 KCNIP1 LCP2 KCNMB1 |
nsv600260 | Chr.5:170265550 - 170532433 on Build GRCh38 | Loss | LINC01366 CTD-2270F17.1 KCNIP1 LCP2 KCNMB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |