Assay Details
Target Gene Details
Entrez Gene ID: | 101927908 |
Gene Name: | uncharacterized LOC101927908 |
Gene Aliases: |
- |
Location: |
Chr.5:167164934-167168401 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927908 | NR_134280.1 |
Target Gene Details
Entrez Gene ID: | 57451 |
Gene Name: | teneurin transmembrane protein 2 |
Gene Aliases: |
ODZ2, TEN-M2, TNM2, ten-2 |
Location: |
Chr.5:166979066-168264159 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TENM2 | XM_005265950.2 | XP_005266007.1 | ||
XM_006714897.2 | XP_006714960.1 | |||
XM_011534604.2 | XP_011532906.1 | |||
XM_017009660.1 | XP_016865149.1 | |||
XM_017009661.1 | XP_016865150.1 | |||
XM_017009662.1 | XP_016865151.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830545 | Chr.5:167120455 - 167225256 on Build GRCh38 | Loss | LOC101927908 TENM2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |