Assay Details
Target Gene Details
Entrez Gene ID: | 1016 |
Gene Name: | cadherin 18 |
Gene Aliases: |
CDH14, CDH14L, CDH24 |
Location: |
Chr.5:19472930-20575902 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1031377 | Chr.5:19289979 - 20971814 on Build GRCh38 | Loss |
![]() ![]() |
nsv597424 | Chr.5:19381946 - 19751051 on Build GRCh38 | Gain |
![]() |
nsv470997 | Chr.5:19441626 - 19534696 on Build GRCh38 | Loss |
![]() |
nsv4752 | Chr.5:19449761 - 19482880 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)