Assay Details
Target Gene Details
Entrez Gene ID: | 4726 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit S6 |
Gene Aliases: |
CI-13kA, CI-13kD-A, CI13KDA |
Location: |
Chr.5:1801382-1816051 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS6 | NM_004553.4 | NP_004544.1 | ||
AF044959.1 | AAC27799.1 | |||
AK022503.1 | ||||
BC038664.1 | AAH38664.1 | |||
BC046155.2 | AAH46155.1 | |||
BG706812.1 | ||||
BM172493.1 | ||||
BM759028.1 | ||||
BM764340.1 | ||||
CN285188.1 | ||||
CR456880.1 | CAG33161.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3603817 | Chr.5:1638442 - 1833414 on Build GRCh38 | Gain | NDUFS6 MRPL36 MIR4277 |
nsv1033147 | Chr.5:1717188 - 1815707 on Build GRCh38 | Gain | NDUFS6 MRPL36 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |