Assay Details
Target Gene Details
Entrez Gene ID: | 9555 |
Gene Name: | H2A histone family member Y |
Gene Aliases: |
H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2 |
Location: |
Chr.5:135334381-135399887 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2AFY | NM_001040158.1 | NP_001035248.1 | ||
NM_004893.2 | NP_004884.1 | |||
NM_138609.2 | NP_613075.1 | |||
NM_138610.2 | NP_613258.2 | |||
XM_005272132.1 | XP_005272189.1 | |||
XM_011543729.2 | XP_011542031.1 | |||
XM_011543730.2 | XP_011542032.1 | |||
XM_011543731.1 | XP_011542033.1 | |||
XM_011543732.1 | XP_011542034.1 | |||
XM_011543733.2 | XP_011542035.1 | |||
XM_017010067.1 | XP_016865556.1 | |||
XM_017010068.1 | XP_016865557.1 | |||
XM_017010069.1 | XP_016865558.1 | |||
AB209490.1 | BAD92727.1 | |||
AF041483.1 | AAC33433.1 | |||
AF044286.1 | AAC33434.1 | |||
AF054174.1 | AAC39908.1 | |||
AK023409.1 | BAB14565.1 | |||
AK123153.1 | 1 | 192 | ||
AK296014.1 | ||||
AY134746.1 | AAN08620.1 | |||
BC013331.1 | AAH13331.1 | |||
BC095406.1 | AAH95406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv599745 | Chr.5:135329392 - 135393177 on Build GRCh38 | Loss | C5orf66 H2AFY |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |