Assay Details
Target Gene Details
Entrez Gene ID: | 9443 |
Gene Name: | mediator complex subunit 7 |
Gene Aliases: |
ARC34, CRSP33, CRSP9 |
Location: |
Chr.5:157138440-157142910 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED7 | NM_001100816.1 | NP_001094286.1 | ||
NM_004270.4 | NP_004261.1 | |||
AF031383.1 | AAC52115.1 | |||
AK308393.1 | ||||
AV711090.1 | ||||
BQ130907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain | FAM71B MED7 ITK HAVCR1 HAVCR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |