Assay Details
Target Gene Details
Entrez Gene ID: | 91942 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 2 |
Gene Aliases: |
B17.2L, MMTN, NDUFA12L, mimitin |
Location: |
Chr.5:60945129-61153037 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF2 | NM_174889.4 | NP_777549.1 | ||
AB183433.1 | BAD91205.1 | |||
AF087990.1 | ||||
AK026051.1 | ||||
AK291296.1 | ||||
BC022869.1 | ||||
BC033965.1 | AAH33965.1 | |||
KU178834.1 | ||||
KU178835.1 | ||||
KU178836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3605201 | Chr.5:60946289 - 60983862 on Build GRCh38 | Loss |
![]() |
nsv1161251 | Chr.5:60943616 - 60988813 on Build GRCh38 | Deletion |
![]() ![]() |
nsv462200 | Chr.5:60840158 - 60971048 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv9810n54 | Chr.5:60820786 - 61017188 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv520871 | Chr.5:60845418 - 61024330 on Build GRCh38 | Loss |
![]() ![]() |
nsv598313 | Chr.5:60951988 - 61079085 on Build GRCh38 | Loss |
![]() |
nsv598308 | Chr.5:60833534 - 61033797 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)