Assay Details
Target Gene Details
Entrez Gene ID: | 8622 |
Gene Name: | phosphodiesterase 8B |
Gene Aliases: |
ADSD, PPNAD3 |
Location: |
Chr.5:77180252-77428256 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE8B | XM_005248623.4 | XP_005248680.1 | ||
XM_005248624.4 | XP_005248681.1 | |||
XM_011543701.2 | XP_011542003.1 | |||
XM_017010007.1 | XP_016865496.1 | |||
XM_017010008.1 | XP_016865497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469515 | Chr.5:77080181 - 77230847 on Build GRCh38 | Loss | PDE8B SNORA47 ZBED3 ZBED3-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |