Assay Details
Target Gene Details
Entrez Gene ID: | 1946 |
Gene Name: | ephrin A5 |
Gene Aliases: |
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS |
Location: |
Chr.5:107376889-107672364 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFNA5 | NM_001962.2 | NP_001953.1 | ||
XM_006714565.2 | XP_006714628.1 | |||
XM_011543250.2 | XP_011541552.1 | |||
XM_011543251.2 | XP_011541553.1 | |||
XM_017009205.1 | XP_016864694.1 | |||
BC075054.2 | AAH75054.1 | |||
BC075055.2 | AAH75055.1 | |||
U26403.1 | AAB60377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34018 | Chr.5:107385211 - 107774805 on Build GRCh38 | Loss |
![]() |
nsv1017067 | Chr.5:106985082 - 108550974 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)