Assay Details
Target Gene Details
Entrez Gene ID: | 54622 |
Gene Name: | ADP ribosylation factor like GTPase 15 |
Gene Aliases: |
ARFRP2 |
Location: |
Chr.5:53883942-54310590 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARL15 | NM_019087.2 | NP_061960.1 | ||
XM_011543498.2 | XP_011541800.1 | |||
XM_011543499.2 | XP_011541801.1 | |||
XM_011543500.2 | XP_011541802.1 | |||
XM_017009598.1 | XP_016865087.1 | |||
AK000058.1 | BAA90915.1 | |||
AK290886.1 | ||||
AK311482.1 | ||||
BC026093.1 | AAH26093.1 | |||
CR457225.1 | CAG33506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5666n100 | Chr.5:53231175 - 54453241 on Build GRCh38 | Gain | NDUFS4 MIR581 LINC01033 ARL15 MIR4459 FST |
nsv830298 | Chr.5:53678082 - 53897568 on Build GRCh38 | Gain | NDUFS4 ARL15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |