Assay Details
Target Gene Details
Entrez Gene ID: | 100505678 |
Gene Name: | STARD4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:111512226-111739726 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD4-AS1 | NR_040093.1 | |||
CR749489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830449 | Chr.5:111414293 - 111585511 on Build GRCh38 | Gain+Loss | CAMK4 STARD4 STARD4-AS1 |