Assay Details
Target Gene Details
Entrez Gene ID: | 10098 |
Gene Name: | tetraspanin 5 |
Gene Aliases: |
NET-4, NET4, TM4SF9, TSPAN-5 |
Location: |
Chr.4:98470367-98658661 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN5 | NM_005723.3 | NP_005714.2 | ||
XM_005262680.1 | XP_005262737.1 | |||
AF065389.1 | AAC17120.1 | |||
AK295385.1 | ||||
AK315723.1 | ||||
BC009704.1 | AAH09704.1 | |||
CR541809.1 | CAG46608.1 | |||
CR541829.1 | CAG46628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1004181 | Chr.4:98287001 - 98608425 on Build GRCh38 | Gain | TSPAN5 RAP1GDS1 |
esv2763801 | Chr.4:98173741 - 98492702 on Build GRCh38 | Gain | TSPAN5 RAP1GDS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |