Assay Details
Target Gene Details
Entrez Gene ID: | 83447 |
Gene Name: | solute carrier family 25 member 31 |
Gene Aliases: |
AAC4, ANT 4, ANT4, SFEC35kDa |
Location: |
Chr.4:127730375-127774299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A31 | NM_001318467.1 | NP_001305396.1 | ||
NM_031291.3 | NP_112581.1 | |||
XM_011532298.1 | XP_011530600.1 | |||
AJ863129.1 | CAI05952.1 | |||
AL136857.1 | CAB66791.1 | |||
AY550240.1 | ||||
BC022032.2 | AAH22032.1 | |||
BG773445.1 | ||||
BM563946.1 | ||||
BQ438347.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830056 | Chr.4:127595984 - 127755402 on Build GRCh38 | Loss | SLC25A31 INTU |
nsv830057 | Chr.4:127670133 - 127867622 on Build GRCh38 | Loss | HSPA4L SLC25A31 INTU |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |