Assay Details
Target Gene Details
Entrez Gene ID: | 7424 |
Gene Name: | vascular endothelial growth factor C |
Gene Aliases: |
Flt4-L, LMPH1D, VRP |
Location: |
Chr.4:176683534-176792745 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VEGFC | NM_005429.4 | NP_005420.1 | ||
AK313879.1 | ||||
BC035212.1 | AAH35212.1 | |||
BC063685.1 | AAH63685.1 | |||
CR541897.1 | CAG46695.1 | |||
U43142.1 | AAA85214.1 | |||
U58111.1 | AAB02909.1 | |||
X94216.1 | CAA63907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion | ASB5 SPCS3 LINC01098 LOC285500 AGA SPATA4 LINC01099 VEGFC GPM6A WDR17 NEIL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |