Assay Details
Target Gene Details
Entrez Gene ID: | 79804 |
Gene Name: | HAND2 antisense RNA 1 (head to head) |
Gene Aliases: |
DEIN, NBLA00301 |
Location: |
Chr.4:173528600-173591255 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAND2-AS1 | NR_136195.1 | |||
NR_136196.1 | ||||
AK054605.1 | ||||
AK123668.1 | ||||
DA863153.1 | ||||
DB292439.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)