Assay Details
Target Gene Details
Entrez Gene ID: | 5602 |
Gene Name: | mitogen-activated protein kinase 10 |
Gene Aliases: |
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK |
Location: |
Chr.4:86012296-86594117 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003585 | Chr.4:86036814 - 86329283 on Build GRCh38 | Loss | MAPK10 LOC101929064 |
dgv5332n100 | Chr.4:86044950 - 86062079 on Build GRCh38 | Loss | MAPK10 |
nsv511259 | Chr.4:86045174 - 86062079 on Build GRCh38 | Loss | MAPK10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |