Assay Details
Target Gene Details
Entrez Gene ID: | 59350 |
Gene Name: | relaxin/insulin like family peptide receptor 1 |
Gene Aliases: |
LGR7, RXFPR1 |
Location: |
Chr.4:158521711-158653372 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830124 | Chr.4:158518665 - 158643187 on Build GRCh38 | Gain |
![]() |
esv3602833 | Chr.4:158437503 - 158589628 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)