Assay Details
Target Gene Details
Entrez Gene ID: | 5156 |
Gene Name: | platelet derived growth factor receptor alpha |
Gene Aliases: |
CD140A, GAS9, PDGFR-2, PDGFR2, RHEPDGFRA |
Location: |
Chr.4:54229097-54298245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDGFRA | NM_006206.4 | NP_006197.1 | ||
XM_005265743.1 | XP_005265800.1 | |||
XM_006714039.3 | XP_006714102.1 | |||
XM_006714041.3 | XP_006714104.1 | |||
XM_017008282.1 | XP_016863771.1 | |||
AK308353.1 | ||||
AK311006.1 | ||||
AK316578.1 | ||||
BC015186.1 | AAH15186.1 | |||
BC063414.1 | AAH63414.1 | |||
DA678599.1 | ||||
M21574.1 | AAA96715.1 | |||
M30494.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002905 | Chr.4:54227510 - 54249531 on Build GRCh38 | Loss | PDGFRA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |