Assay Details
Target Gene Details
Entrez Gene ID: | 306 |
Gene Name: | annexin A3 |
Gene Aliases: |
ANX3 |
Location: |
Chr.4:78551588-78610451 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANXA3 | NM_005139.2 | NP_005130.1 | ||
AK313945.1 | ||||
BC000871.1 | AAH00871.1 | |||
BX336217.2 | ||||
M20560.1 | AAA59496.1 | |||
M63310.1 | AAA52284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1006026 | Chr.4:78111837 - 79966019 on Build GRCh38 | Loss | NAA11 MIR5096 BMP2K LOC101928893 FRAS1 LINC00989 ANXA3 PAQR3 LINC01088 GK2 ANTXR2 LINC01094 PCAT4 |
nsv1009633 | Chr.4:77966419 - 78731082 on Build GRCh38 | Gain | LOC101928893 FRAS1 LINC01094 ANXA3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |