Assay Details
Target Gene Details
Entrez Gene ID: | 353497 |
Gene Name: | DNA polymerase nu |
Gene Aliases: |
POL4P |
Location: |
Chr.4:2071958-2242164 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLN | NM_181808.3 | NP_861524.2 | ||
AF044578.1 | AAD02338.1 | |||
AK131239.1 | BAD18421.1 | |||
AY136549.1 | AAN52116.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion | NAT8L ADD1 MFSD10 SH3BP2 CFAP99 FAM193A TNIP2 ZFYVE28 MXD4 MIR4800 RNF4 NOP14-AS1 C4orf48 HAUS3 NOP14 POLN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |