Assay Details
Target Gene Details
Entrez Gene ID: | 113510 |
Gene Name: | helicase, POLQ-like |
Gene Aliases: |
HEL308 |
Location: |
Chr.4:83407343-83455883 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HELQ | NM_001297755.1 | NP_001284684.1 | ||
NM_001297756.1 | NP_001284685.1 | |||
NM_001297757.1 | NP_001284686.1 | |||
NM_133636.3 | NP_598375.2 | |||
XM_005262711.1 | XP_005262768.1 | |||
XM_005262713.2 | XP_005262770.1 | |||
XM_006714076.2 | XP_006714139.1 | |||
XM_017007679.1 | XP_016863168.1 | |||
XM_017007680.1 | XP_016863169.1 | |||
XM_017007682.1 | XP_016863171.1 | |||
XM_017007683.1 | XP_016863172.1 | |||
AF436845.1 | AAL85274.1 | |||
AK308395.1 | ||||
AK312725.1 | ||||
AL512702.1 | CAH56359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893971 | Chr.4:83096792 - 83493129 on Build GRCh38 | Gain | FAM175A PLAC8 HPSE COQ2 HELQ MRPS18C |
esv3601196 | Chr.4:83419958 - 83470239 on Build GRCh38 | Loss | FAM175A HELQ MRPS18C |
nsv829992 | Chr.4:83244588 - 83458578 on Build GRCh38 | Loss | HPSE COQ2 HELQ MRPS18C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |