Assay Details
Target Gene Details
Entrez Gene ID: | 55714 |
Gene Name: | teneurin transmembrane protein 3 |
Gene Aliases: |
MCOPCB9, ODZ3, TNM3, Ten-m3, ten-3 |
Location: |
Chr.4:181447613-182803024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TENM3 | XM_017008385.1 | XP_016863874.1 | ||
XM_017008389.1 | XP_016863878.1 | |||
XM_017008390.1 | XP_016863879.1 | |||
XM_017008391.1 | XP_016863880.1 | |||
XM_017008392.1 | XP_016863881.1 | |||
XM_017008393.1 | XP_016863882.1 | |||
XM_017008394.1 | XP_016863883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830163 | Chr.4:181391765 - 181555717 on Build GRCh38 | Gain |
![]() |
nsv596307 | Chr.4:181379580 - 181556233 on Build GRCh38 | Gain |
![]() |
nsv1021069 | Chr.4:181273983 - 181566272 on Build GRCh38 | Loss |
![]() |
nsv1016288 | Chr.4:179729085 - 182726257 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)