Assay Details
Target Gene Details
Entrez Gene ID: | 25945 |
Gene Name: | nectin cell adhesion molecule 3 |
Gene Aliases: |
CD113, CDW113, NECTIN-3, PPR3, PRR3, PVRL3, PVRR3 |
Location: |
Chr.3:111071759-111201444 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NECTIN3 | NM_001243286.1 | NP_001230215.1 | ||
NM_001243288.1 | NP_001230217.1 | |||
NM_015480.2 | NP_056295.1 | |||
XM_011512663.1 | XP_011510965.1 | |||
XM_017006123.1 | XP_016861612.1 | |||
XM_017006124.1 | XP_016861613.1 | |||
XM_017006125.1 | XP_016861614.1 | |||
XM_017006126.1 | XP_016861615.1 | |||
XM_017006127.1 | XP_016861616.1 | |||
AF282874.1 | AAF97597.1 | |||
AK075105.1 | BAC11404.1 | |||
BC067808.1 | AAH67808.1 | |||
BI464010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460821 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain | CD96 PHLDB2 PLCXD2 PLCXD2-AS1 ZBED2 PVRL3-AS1 LOC151760 NECTIN3 |
nsv591276 | Chr.3:111103649 - 111135349 on Build GRCh38 | Loss | NECTIN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |