Assay Details
Target Gene Details
Entrez Gene ID: | 64091 |
Gene Name: | popeye domain containing 2 |
Gene Aliases: |
POP2 |
Location: |
Chr.3:119642052-119660590 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POPDC2 | NM_001308333.1 | NP_001295262.1 | ||
NM_022135.3 | NP_071418.2 | |||
XM_005247698.3 | XP_005247755.1 | |||
AF204173.1 | AAG23406.1 | |||
AK226022.1 | ||||
AK314819.1 | ||||
AK377174.1 | ||||
AK377176.1 | ||||
AK377177.1 | ||||
AK377178.1 | ||||
AK377179.1 | ||||
AK377180.1 | ||||
AK377181.1 | ||||
AK377182.1 | ||||
AK377183.1 | ||||
AK377184.1 | ||||
BC026911.2 | AAH26911.1 | |||
BC044929.1 | AAH44929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014394 | Chr.3:119641659 - 119693428 on Build GRCh38 | Loss | POPDC2 COX17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |