Assay Details
Target Gene Details
Entrez Gene ID: | 80321 |
Gene Name: | centrosomal protein 70 |
Gene Aliases: |
BITE |
Location: |
Chr.3:138494339-138594383 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv999041 | Chr.3:138429970 - 138629146 on Build GRCh38 | Gain | CEP70 FAIM ESYT3 |
nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss | DBR1 PIK3CB MRAS ESYT3 LINC01210 DZIP1L CEP70 FAIM CLDN18 A4GNT SOX14 NME9 ARMC8 |
esv3597916 | Chr.3:138478438 - 138638584 on Build GRCh38 | Gain | CEP70 FAIM ESYT3 |
dgv4908n100 | Chr.3:138494324 - 139018846 on Build GRCh38 | Gain | FOXL2 CEP70 PIK3CB LINC01391 PRR23A FAIM FOXL2NB |
nsv522517 | Chr.3:138455681 - 138579140 on Build GRCh38 | Loss | CEP70 ESYT3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |