Assay Details
Target Gene Details
Entrez Gene ID: | 10267 |
Gene Name: | receptor activity modifying protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:237858714-237912117 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAMP1 | NM_001308353.1 | NP_001295282.1 | ||
NM_005855.3 | NP_005846.1 | |||
XM_017003152.1 | XP_016858641.1 | |||
XM_017003153.1 | XP_016858642.1 | |||
XM_017003155.1 | XP_016858644.1 | |||
XM_017003156.1 | XP_016858645.1 | |||
AI017108.1 | ||||
AJ001014.1 | CAA04472.1 | |||
AY265457.1 | AAP23298.1 | |||
BC000548.2 | AAH00548.1 | |||
BI548986.1 | ||||
CR542032.1 | CAG46829.1 | |||
CR542044.1 | CAG46841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv584754 | Chr.2:237865062 - 237894836 on Build GRCh38 | Loss | RAMP1 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | ERFE LOC151174 UBE2F ESPNL RBM44 COPS8 RAB17 LINC01107 HES6 PER2 LOC643387 ASB1 TRAF3IP1 SCLY MIR6811 ACKR3 COL6A3 AGAP1 MLPH LOC93463 GBX2 LOC105373991 IQCA1 LOC105373944 LOC105373958 RAMP1 ASB18 KLHL30 ILKAP LRRFIP1 PRLH UBE2F-SCLY LOC101927958 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |