Assay Details
Target Gene Details
Entrez Gene ID: | 9874 |
Gene Name: | tousled like kinase 1 |
Gene Aliases: |
PKU-beta |
Location: |
Chr.2:170990823-171231314 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TLK1 | NM_001136554.1 | NP_001130026.1 | ||
NM_001136555.1 | NP_001130027.1 | |||
NM_012290.4 | NP_036422.3 | |||
XM_011512238.2 | XP_011510540.1 | |||
XM_017005421.1 | XP_016860910.1 | |||
XM_017005422.1 | XP_016860911.1 | |||
AB004885.1 | BAA20562.1 | |||
AF162666.1 | AAF03094.1 | |||
AF246219.1 | AAF71263.1 | |||
AK090779.1 | ||||
AK292508.1 | ||||
AK301857.1 | ||||
BC032657.1 | AAH32657.1 | |||
D50927.2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |