Assay Details
Target Gene Details
Entrez Gene ID: | 2281 |
Gene Name: | FK506 binding protein 1B |
Gene Aliases: |
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase |
Location: |
Chr.2:24033205-24067743 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP1B | NM_001322963.1 | NP_001309892.1 | ||
NM_001322964.1 | NP_001309893.1 | |||
NM_004116.4 | NP_004107.1 | |||
NM_054033.3 | NP_473374.1 | |||
NR_136536.1 | ||||
NR_136538.1 | ||||
NR_136539.1 | ||||
XM_011532699.2 | XP_011531001.1 | |||
XM_017003593.1 | XP_016859082.1 | |||
XM_017003594.1 | XP_016859083.1 | |||
XM_017003596.1 | XP_016859085.1 | |||
AF322070.1 | AAK11191.1 | |||
BC002614.1 | AAH02614.1 | |||
BF216711.1 | ||||
BF969580.1 | ||||
BI546013.1 | ||||
BM802489.1 | ||||
BU147063.1 | ||||
BU508033.1 | ||||
D38037.1 | BAA07232.1 | |||
L37086.1 | AAC37581.1 | |||
S69800.1 | AAB30685.1 | |||
S69815.1 | AAB30684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain | LOC105374325 SF3B6 FAM228A TP53I3 ITSN2 PFN4 FAM228B UBXN2A KLHL29 MFSD2B FKBP1B WDCP ATAD2B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |