Assay Details
Target Gene Details
Entrez Gene ID: | 56888 |
Gene Name: | potassium channel modulatory factor 1 |
Gene Aliases: |
DEBT91, FIGC, PCMF, ZZZ1 |
Location: |
Chr.2:84971108-85059472 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCMF1 | NM_020122.4 | NP_064507.3 | ||
XM_006712052.3 | XP_006712115.1 | |||
XM_011532990.1 | XP_011531292.1 | |||
AB083199.1 | BAC43745.1 | |||
AF155652.1 | AAF67009.1 | |||
AK023403.1 | BAB14563.1 | |||
AK314761.1 | ||||
BC000178.2 | AAH00178.1 | |||
BU844982.1 | ||||
CT004343.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)