Assay Details
Target Gene Details
Entrez Gene ID: | 80895 |
Gene Name: | ILK associated serine/threonine phosphatase |
Gene Aliases: |
ILKAP2, ILKAP3, PP2C-DELTA, PPM1O |
Location: |
Chr.2:238170401-238203702 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ILKAP | NM_030768.2 | NP_110395.1 | ||
XM_005246106.1 | XP_005246163.1 | |||
XM_006712784.1 | XP_006712847.1 | |||
XM_011511946.2 | XP_011510248.1 | |||
XM_017005056.1 | XP_016860545.1 | |||
XM_017005057.1 | XP_016860546.1 | |||
XM_017005058.1 | XP_016860547.1 | |||
AK001043.1 | ||||
AK055417.1 | ||||
AL136850.1 | CAB66784.1 | |||
AY024365.1 | AAK07736.1 | |||
BC006576.2 | AAH06576.1 | |||
CR533533.1 | CAG38564.1 | |||
FJ491380.1 | ||||
HQ658059.1 | ||||
HQ658060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | LOC105373991 LINC01107 LOC151174 ASB18 LOC105373958 LOC101927958 AGAP1 TRAF3IP1 MIR6811 ASB1 GBX2 UBE2F SCLY COPS8 LOC643387 ILKAP ERFE COL6A3 UBE2F-SCLY LOC105373944 ACKR3 HES6 KLHL30 RAMP1 RBM44 MLPH IQCA1 PER2 RAB17 PRLH LOC93463 ESPNL LRRFIP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |