Assay Details
Target Gene Details
Entrez Gene ID: | 57504 |
Gene Name: | metastasis associated 1 family member 3 |
Gene Aliases: |
- |
Location: |
Chr.2:42494569-42795032 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTA3 | NM_001282755.1 | NP_001269684.1 | ||
XM_005264459.2 | XP_005264516.1 | |||
AK127245.1 | ||||
DA239202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3583763 | Chr.2:42479558 - 42513741 on Build GRCh38 | Loss | MTA3 KCNG3 |
nsv524679 | Chr.2:42350030 - 42576225 on Build GRCh38 | Loss | MTA3 COX7A2L KCNG3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |