Assay Details
Target Gene Details
Entrez Gene ID: | 10267 |
Gene Name: | receptor activity modifying protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:237858714-237912117 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAMP1 | NM_001308353.1 | NP_001295282.1 | ||
NM_005855.3 | NP_005846.1 | |||
XM_017003152.1 | XP_016858641.1 | |||
XM_017003153.1 | XP_016858642.1 | |||
XM_017003154.1 | XP_016858643.1 | |||
XM_017003156.1 | XP_016858645.1 | |||
AI017108.1 | ||||
AJ001014.1 | CAA04472.1 | |||
AY265457.1 | AAP23298.1 | |||
BC000548.2 | AAH00548.1 | |||
BI548986.1 | ||||
BM978161.1 | ||||
CR542032.1 | CAG46829.1 | |||
CR542044.1 | CAG46841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv584754 | Chr.2:237865062 - 237894836 on Build GRCh38 | Loss | RAMP1 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | LOC101927958 LOC151174 LOC105373991 LINC01107 RAMP1 PER2 LOC93463 ASB18 UBE2F PRLH LRRFIP1 IQCA1 HES6 LOC105373958 TRAF3IP1 COPS8 ASB1 GBX2 COL6A3 LOC105373944 SCLY MLPH ESPNL ERFE ILKAP MIR6811 AGAP1 UBE2F-SCLY RAB17 LOC643387 RBM44 ACKR3 KLHL30 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |